Cole-Parmer® Handheld Ultraviolet Lamps
Portable, lightweight, easy to use
  • High UV intensity gets the job done
  • Indicator lights alert you if the battery is low, or normal strength
The series of lightweight, handheld lamps offer UV wavelengths of 365 and 254 nm, or a combination of the two. They’re cordless and use 12 VDC rechargeable batteries. They can sustain three hours of continuous use or about 500 intermittent uses when fully charged. The models equipped with an automatic safety switch prevent harmful exposure to UV radiation if the lamp is turned upside down.
44,300.00 - ₹71,000.00INR / Each
19 variations of this product are available.
See All Product Options Below
19 PRODUCT OPTIONS
Compareitem specifications
Power (VAC) Wavelength Description
Availability Pricing
COMPARE
110 254 Handheld UV Lamp, 6 Watt, 254 nm, AC Operated; 110 VAC, 50/60 Hz
₹44,300.00INR / Each
COMPARE
110 365 Handheld UV Lamp, 6 Watt, 365 nm, AC Operated; 110 VAC, 50/60 Hz
₹44,300.00INR / Each
COMPARE
110 254/365 Handheld UV Lamp, 6 Watt, 254/365 nm, AC Operated; 110 VAC, 50/60 Hz
₹44,300.00INR / Each
COMPARE
110 254 Handheld UV Lamp with Safety Switch, 6 Watt, 254 nm, Rechargeable Battery; 110 VAC
₹49,800.00INR / Each
COMPARE
110 365 Handheld UV Lamp with Safety Switch, 6 Watt, 365 nm, Rechargeable Battery; 110 VAC
₹49,800.00INR / Each
COMPARE
110 254/365 Handheld UV Lamp with Safety Switch, 6 Watt, 254/365 nm, Rechargeable Battery; 110 VAC
₹49,800.00INR / Each
COMPARE
110 254 Handheld UV Lamp, 6 Watt, 254 nm, Rechargeable Battery; 110 VAC
₹49,600.00INR / Each
COMPARE
110 365 Handheld UV Lamp, 6 Watt, 365 nm, Rechargeable Battery; 110 VAC
₹48,900.00INR / Each
COMPARE
110 254/365 Handheld UV Lamp, 6 Watt, 254/365 nm, Rechargeable Battery; 110 VAC
₹48,900.00INR / Each
COMPARE
110 / 240 254 Handheld UV Lamp with Safety Switch, 6 Watt, 254 nm, Rechargeable Battery; 110 to 240 VAC
₹52,600.00INR / Each
COMPARE
110 / 240 365 Handheld UV Lamp with Safety Switch, 6 Watt, 365 nm, Rechargeable Battery; 110 to 240 VAC
₹52,600.00INR / Each
COMPARE
110 / 240 254/365 Handheld UV Lamp with Safety Switch, 6 Watt, 254/365 nm, Rechargeable Battery; 110 to 240 VAC
₹52,600.00INR / Each
COMPARE
110 / 240 254 Handheld UV Lamp, 6 Watt, 254 nm, Rechargeable Battery; 110 to 240 VAC
₹51,700.00INR / Each
COMPARE
110 / 240 365 Handheld UV Lamp, 6 Watt, 365 nm, Rechargeable Battery; 110 to 240 VAC
₹51,700.00INR / Each
COMPARE
110 / 240 254/365 Handheld UV Lamp, 6 Watt, 254/365 nm, Rechargeable Battery; 110 to 240 VAC
₹51,700.00INR / Each
COMPARE
220 365 Handheld UV Lamp, 6 Watt, 365 nm, AC Operated; 220 VAC, 50 Hz UK
₹44,300.00INR / Each
COMPARE
220 254/365 Handheld UV Lamp, 6 Watt, 254/365 nm, AC Operated; 220 VAC, 50 Hz UK
₹44,300.00INR / Each
COMPARE
220 254/365 Handheld UV Lamp, 6 Watt, 254/365 nm, AC Operated; 220 VAC, 50 Hz SA
₹44,300.00INR / Each
COMPARE
220 254/365 Handheld UV Lamp, 6 Watt, 254/365 nm, AC Operated; 220 VAC, 50 Hz Euro
₹71,000.00INR / Each
Showing 1- 19 of 19