Cole-Parmer® FJT-200 Series Stuart Flocculation Jar Testers
Get repeatable conditions between samples
  • Easily view paddle speed on digital display
  • Save time with two preset programs
  • Built-in illumination and dark background for easy viewing of samples
Achieve repeatable conditions between samples and from run to run with these flocculation jar testers. Speed control with electronic feedback accurately maintains the rotational speed of the stainless steel paddles. Two preset programs allow routine speed and time parameters to be entered, stored in memory, and recalled or altered at any time.
Select two-position tester 99551-00 with a handle for easy portability; six-position tester 99551-05 with touchpad control features a digital timer. Both testers accept 1-L beakers (not included).
268,000.00 - ₹389,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
No. Of Stirring Positions Timer Description
Availability Pricing
COMPARE
2 No Flocculation Jar Tester, 2-place; 230 VAC, 50/60 Hz
₹268,000.00INR / Each
COMPARE
6 Yes Flocculation Jar Tester, 6-place; 230 VAC, 50 Hz
₹389,000.00INR / Each