Cole-Parmer® WS-200 Series Stuart Water Stills
Get high purity water with all-glass construction
  • Two independent safety thermostats prevent overheating
  • Heating element shuts down when reservoir is full
This all-glass water still with silica-sheathed heaters and high-efficiency condenser ensures top distillate purity at an economical price. A reservoir level control feature automatically turns off the heater when the reservoir is full preventing any overflow. Two independent safety thermostats, mounted in the condenser and boiler, prevent overheating in the event the water supply is interrupted.
Easily and safely change the hoses without the risk of breaking any glassware using the screw thread connections. Built-in acid addition funnel and drain stopcock allow for easy descaling without dismantling the glassware. Use the still on a benchtop or wall mount using the two predrilled screw holes in the stand.
264,000.00 - ₹265,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Description Capacity Power (VAC)
Availability Pricing
COMPARE
Distinction Water Still, 240 VAC 4 L/hr 240
₹265,000.00INR / Each
COMPARE
Distinction Water Still, 220 VAC 4 L/hr 220
₹264,000.00INR / Each