This product is now discontinued, View Related Products
Stuart Bibbyjet Pro Pipette Controller
Built-in non-return valve prevents liquid contamination
  • Ergonomic design allows for fatigue-free operation
  • Provides up to 8 hours of full speed operation on a single charge
  • Accepts glass or plastic serological pipettes from 0.1 to 200 mL
  • Removable and autoclavable silicone adapter, filter, and nosepiece
The lightweight, ergonomic pipette controller features two main function buttons for complete control over filling and delivery. An integrated speed control dial allows you to adjust continuously how slowly or quickly you want to take up liquid, while a simple flick of a switch changes the dispensing method from power assisted to non-power assisted.
Built-in non-return valve combined with a membrane filter protects against penetration of liquids. To protect the controller from internal corrosion, vapors are expelled from the front of the unit, away from the operator and not through the pump. The silicone adapter included with the controller provides a tight fit for all graduated and volumetric glass or plastic pipettes in the volume range of 0.1 mL to 200 mL. The adapter along with the filter and nosepiece are autoclavable at 250°F (121°C).
A rechargeable NiMh battery provides up to 8 hours of nonstop pipetting on a 4-hour charge. LED indicator flashes when 2 hours of pipetting time remain.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Volume (mL) Description
Availability Pricing
COMPARE
200 UK plug
Call For Price
COMPARE
200 EU plug