This product is now discontinued, View Related Products
Cole-Parmer Stuart Analog Hot Plates
Custom designed hot plates provide accurate temperature control
  • Hot surface warning light flashes when plate temperature is above 50°C
  • Durable front panel reists spills and chemicals
  • Available in various plate surface and body color configurations
Ideal for day-to-day use by scientific professionals and students, the Stuart analog hot plates offer precise temperature control of your samples. A ‘hockey stick’ LED array displays the set and actual temperature of the unit. The surface temperature range is indicated by progressively illuminating LED lights. To achieve accurate sample temperature control up to 200°C, a digital temperature controller (sold separately) can be connected to the rear of the unit.
The simple body design features a spill-resistant front control panel with chemically toughened glass for extra resilience. When the plate temperature rises above 50°C, a hot warning light flashes even if the unit is turned off and unplugged. All units features an integral fixing point for a retort rod and BioCote™ antimicrobial protection.
Each hot plate is custom designed. Choose between a glass ceramic or ceramic-coated aluminum plate surface and select a favorite body color—blue, black, or white. The glass ceramic plate is chemical resistant, easy to clean, and allows for very high plate temperatures; plus the white surface ensures good visibility of color changes. The ceramic-coated aluminum plate offers excellent heat transmission, rapid heating with a 700-watt element, even temperature distribution, and is chemical resistant.
93,500.00 - ₹103,000.00INR / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° F) Power (VAC) Description
Availability Pricing
COMPARE
617 120 Analog Hot Plate, Aluminum, Blue, 120V
Call For Price
COMPARE
617 230 Analog Hot Plate, Aluminum, Blue, 230V
₹93,500.00INR / Each
COMPARE
617 120 Analog Hot Plate, Aluminum, White, 120V
Call For Price
COMPARE
617 230 Analog Hot Plate, Aluminum, White, 230V
₹94,400.00INR / Each
COMPARE
617 120 Analog Hot Plate, Aluminum, Black, 120V
Call For Price
COMPARE
617 230 Analog Hot Plate, Aluminum, Black, 230V
₹94,400.00INR / Each
COMPARE
842 120 Analog Hot Plate, Ceramic, Blue, 120V
Call For Price
COMPARE
842 230 Analog Hot Plate, Ceramic, Blue, 230V
₹103,000.00INR / Each
COMPARE
842 120 Analog Hot Plate, Ceramic, White, 120V
Call For Price
COMPARE
842 230 Analog Hot Plate, Ceramic, White, 230V
₹103,000.00INR / Each
COMPARE
842 120 Analog Hot Plate, Ceramic, Black, 120V
Call For Price
COMPARE
842 230 Analog Hot Plate, Ceramic, Black, 230V
₹103,000.00INR / Each
Showing 1- 12 of 12