This product is now discontinued, View Related Products
Stuart Analog Ceramic-Top Hot Plates
Easily control temperature with a dial
  • Heat one large vessel or several small ones
  • Be safe—flashing light warns when top plate is hot
Hotplate temperature is controlled by an easy to use dial. Each model features a "Hot" warning light which flashes whenever the plate temperature is above 50°C. Chemical-resistant ceramic stays cooler around the edges to help prevent accidental burns.
108,000.00 - ₹174,000.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Power (VAC) Top Plate Length (in) Top Plate Material
Availability Pricing
COMPARE
230 12 Ceramic
₹116,000.00INR / Each
COMPARE
120 12 Ceramic
₹159,000.00INR / Each
COMPARE
230 20 Ceramic
₹174,000.00INR / Each
COMPARE
₹108,000.00INR / Each
COMPARE
₹111,000.00INR / Each
COMPARE
₹173,000.00INR / Each
COMPARE
₹174,000.00INR / Each
Showing 1- 7 of 7