This product is now discontinued, View Related Products
Stuart Aluminum-Top Analog Hot Plates
Easily control temperature with a dial
  • Heat one large vessel or several small ones
  • Be safe—flashing light warns when top plate is hot
Hotplate temperature is controlled by an easy to use dial. Each model features a "Hot" warning light which flashes whenever the plate temperature is above 50°C. Rugged aluminum heats quickly and has excellent conduction.
117,000.00 - ₹148,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Power (VAC) Top Plate Length (in)
Availability Pricing
COMPARE
230 20
₹148,000.00INR / Each
COMPARE
230 12
₹117,000.00INR / Each
COMPARE
120 12