This product is now discontinued, View Related Products
Stuart Advanced Analog Stirring Hot Plate with FREE Temperature Controller
Simple and economical
  • Compact, space-saving design suits even cluttered laboratories
  • Powerful magnets provide strong stirring action
  • Be safe—flashing light warns when top plate is hot, and safety circuit protects against overheating
  • Colored LED indicator scale displays approximate plate temperature
These general purpose stirring hot plates are designed with safety as well as performance in mind. The compact shape takes up less bench space and makes storage easier. Powerful magnets and motor give stirring speeds up to 2000 rpm and is capable of mixing large volumes (up to 15 L*). All models include an integral fitting for attaching a support rod.
Advanced stirring hot plates feature either coated aluminum top plate for rapid heating and better temperature uniformity or chemically resistant glass ceramic that stays cool around the edges to help prevent burns. The "Hot" warning light will flash whenever the plate temperature is above 50°C even when the hotplate is turned off (lasts up to 30 minutes without power).
Added to the standard features is an innovative LED temperature indicator scale, which lights up in colored increments as the top plate heats up and shows you when the plate has reached its temperature set point. Use in conjunction with the digital temperature controller (included for FREE) to accurately regulate sample temperature.
*Based on water contained in a 20 L glass-bottom flask
113,000.00 - ₹206,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Advanced Stirring Hot Plate & Free Temperature Controller, Ceramic, 120 V
₹206,000.00INR / Each
COMPARE
Advanced Stirring Hot Plate & Free Temperature Controller, Aluminum, 120 V
₹113,000.00INR / Each