This product is now discontinued, View Related Products
Stuart 3D Gyratory Rocker/Shaker with FREE Tier System
Small space saving design
  • Digital speed control and built-in timer
  • Tier system increases capacity
These rocking shakers provide a 3D gyratory motion—ideal for low foaming agitation, DNA extractions, and staining/de-staining procedures. The angle of tilt can be moved to any position by hand to optimize mixing of vessels. Digital selection of both speed and time facilitates accurate and reproducible conditions; both parameters are controlled through a control knob.
197,000.00 - ₹219,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Mini Gyratory Rocking Shaker with Free Tier System, 120 VAC
₹219,000.00INR / Each
COMPARE
Mini See-Saw Rocking Shaker with Free Tier System, 120 VAC
₹197,000.00INR / Each