This product is now discontinued, View Related Products
Stir-Pak Heavy-Duty Mixers Complete Systems
Modular system for maximum versatility
  • Easily customize your mixer to suit your requirements
Our modular Stir-Pak® heavy-duty mixer system allows you the flexibility to create the best system for your mixing applications. System components are sold separately, and as convenient, complete system packages—customize your own system or select one of ours.
To customize your own system, simply start with a solid-state speed controller, add one of the four mixing heads depending on your speed and torque requirements, select a shaft and propeller, then choose from clamps and stands that best suit your application.
Call For Price
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Max Speed (rpm) Max Torque (in-oz) Power (VAC)
Availability Pricing
COMPARE
180 340 115
Call For Price
COMPARE
180 340 230
Call For Price
COMPARE
900 70 115
Call For Price
COMPARE
2300 70 230
Call For Price
COMPARE
2300 45 115
Call For Price
COMPARE
2300 45 230
Call For Price
COMPARE
6000 17 115
Call For Price
COMPARE
6000 17 230
Call For Price
Showing 1- 8 of 8