This product is now discontinued, View Related Products
Stir-Pak Heavy-Duty Mixer Heads
Modular system for maximum versatility - order components separately
These rugged mixer heads feature TENV, permanent magnet DC motor with permanently lubricated ball bearing construction. Choose from four models depending on your desired speed and torque ranges. All mixer heads are equipped with an integral adjustable-tilt mounting assembly that permits the rotation of the mixer head from 0 to 30° for proper placement of shaft and propeller. Heads measure approximately 11-1⁄2"H x 3-5⁄8" dia (292 mm H x 92 mm dia). Mixer heads are IP24 rated.
Note: Integrated safety switch atop all mixer heads for on/off control closest to mixing action.
High-Torque, Low-Speed Mixer Heads feature a keyless chuck that accepts mixing shafts from 5⁄16" to 3⁄8" (8 to 10 mm) dia.
High-Speed, Low-Torque Mixer Heads incorporate a hollow through-shaft and a precision hand-tightened collet; collet accepts 3⁄8" (10 mm) dia mixing shafts. The design lets you quickly position the mixing propeller at any depth within the container without moving the mixer head. The design also minimizes shaft wobble common at higher speeds and with longer shaft lengths.
Call For Price
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Max Speed (rpm) Max Torque (in-oz) Power (VAC)
Availability Pricing
COMPARE
180 340 115
Call For Price
COMPARE
180 340 230
Call For Price
COMPARE
900 70 115
Call For Price
COMPARE
900 70 230
Call For Price
COMPARE
2300 45 115
Call For Price
COMPARE
2300 45 230
Call For Price
COMPARE
6000 17 115
Call For Price
COMPARE
6000 17 230
Call For Price
Showing 1- 8 of 8