Sterilizable Packaging Material - BOP Reels
BOP Reels materials exhibit total and uniform peelability characteristics with minimal fibre lift, irrespective of speed or conditions of opening. The BOP range of products will eliminate reprocessing costs and equally help in reducing nosocomial infection, and by the ease of use ultimately increase patient safety.
Technical Characteristics:
Non Tearing and Shaterless Film: A unique Film specifically formulated to eliminate tearing, a major weakness in standard materials. The resistance to tear is some 10 times greater and virtually prevents any shattering on seals when opening. Paper: The paper used for BOP reels incorporates an adopted surface treatment which not only allows uniform and constant heat sealing, but will also reduce considerably the shedding of fibre when opened.
Indicat
Call For Price
20 variations of this product are available.
See All Product Options Below
20 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Width (in)
Availability Pricing
COMPARE
2
Call For Price
COMPARE
3
Call For Price
COMPARE
4
Call For Price
COMPARE
5
Call For Price
COMPARE
6
Call For Price
COMPARE
8
Call For Price
COMPARE
10
Call For Price
COMPARE
12
Call For Price
COMPARE
15
Call For Price
COMPARE
17
Call For Price
COMPARE
2
Call For Price
COMPARE
3
Call For Price
COMPARE
4
Call For Price
COMPARE
5
Call For Price
COMPARE
6
Call For Price
COMPARE
8
Call For Price
COMPARE
10
Call For Price
COMPARE
12
Call For Price
COMPARE
15
Call For Price
COMPARE
16
Call For Price
Showing 1- 20 of 20