Stainless Steel Generator Probes for Cole-Parmer® HO-600 Homogenizers
Easily handles fine to medium samples
  • Resistant to most organic solvents
  • Easy to disassemble and clean
These stainless-steel generator probes feature a stator design which allows fine to medium samples to pass through the processing windows. The probes can be easily disassembled for cleaning and are available with PTFE lower and stainless-steel upper bearings.
Compatiable with Cole-Parmer HO-600 homgenizers.
257,000.00 - ₹283,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Max Sample Size (mL) Min Sample Size (mL)
Availability Pricing
COMPARE
316 Stainless Steel 5 0.2
₹257,000.00INR / Each
COMPARE
316 Stainless Steel 5 0.2
₹257,000.00INR / Each
COMPARE
316 Stainless Steel 100 1.5
₹283,000.00INR / Each