This product is now discontinued, View Related Products
StableTemp Muffle Furnaces with Single Set Point Control
Reach maximum temperature quickly—rise times as fast as 25 minutes
  • Autotune feature helps eliminate temperature overshoot
  • Energy efficient insulation keeps exterior safe to touch
These furnaces are available in three sizes to meet your application requirements. All models have autotune feature which allows the furnace to perform better at a particular set point and load, eliminating temperature overshoot for a commonly used application.
The fully embedded heating elements maintain temperature uniformity. A door switch cuts power to the heating elements when the door is opened minimizing your exposure to hot temperatures and extending the life of the heating elements. Unique ceramic fiber insulation and double walled construction minimize exterior surface temperatures. The removable hearth plate prevents damage to
furnace from spills.
Single Set Point Control Furnaces have two LED displays to simultaneously display actual and set point temperatures.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Capacity (Liters) Power (VAC)
Availability Pricing
COMPARE
1.6 120
Call For Price
COMPARE
5.3 240
Call For Price
COMPARE
18.4 208 / 240
Call For Price
COMPARE
42.5 208 / 240