This product is now discontinued, View Related Products
StableTemp Muffle Furnaces with Basic Control
Reach maximum temperature quickly—rise times as fast as 25 minutes
  • Autotune feature helps eliminate temperature overshoot
  • Energy efficient insulation keeps exterior safe to touch
These model CBF furnaces are available in three sizes. All models have autotune feature which allows the furnace to perform better at a particular set point and load, eliminating temperature overshoot for a commonly used application.
The fully embedded heating elements maintain temperature uniformity. A door switch cuts power to the heating elements when the door is opened minimizing your exposure to hot temperatures and extending the life of the heating elements. Unique ceramic fiber insulation and double walled construction minimize exterior surface temperatures. The removable hearth plate prevents damage to
furnace from spills.
Basic Control Furnaces feature a single LED display that toggles between actual and set point temperatures.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Capacity (Liters) Power (VAC)
Availability Pricing
COMPARE
5.3 120
Call For Price
COMPARE
5.3 240
Call For Price
COMPARE
18.4 208 / 240
Call For Price
COMPARE
42.5 208 / 240