This product is now discontinued, View Related Products
StableTemp Chromatography Refrigerators
Performance and security required for everyday chromatography applications
Designed for the chromatography lab, these refrigerators feature two 2" (5.1 cm) dia capped access portholes in each side wall to permit the insertion of thermocouples and other wiring without disturbing temperature settings. Push-button lights and circuit breaker-protected duplex electrical receptacles simplify equipment operation and allow observation of ongoing separations.
Units feature microprocessor-controlled thermostats and digital temperature display for precise temperature control, and thick polyurethane foam insulation. Painted steel exterior and white interior provide durability and easy cleanup. Doors incorporate double-paned argon-filled glass to improve efficiency.
Other features include keyed power switch, chart recorder (select models), high/low temperature alarm, door ajar alarm (except on sliding glass doors), and automatic defrost.
Call For Price
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Number of Doors Refrigerator Capacity (cu ft) Description
Availability Pricing
COMPARE
1 27 Chromatography refrigerator; 27 cu ft, 115V
Call For Price
COMPARE
1 27 Chromatography refrigerator; 27 cu ft, with chart recorder, 115V
Call For Price
COMPARE
2 38 Chromatography refrigerator; 38 cu ft, 115V
Call For Price
COMPARE
2 38 Chromatography refrigerator; 38 cu ft, with chart recorder, 115V
Call For Price
COMPARE
2 45 Chromatography refrigerator; 45 cu ft, 115V
Call For Price
COMPARE
2 45 Chromatography refrigerator; 45 cu ft, with chart recorder, 115V
Call For Price
COMPARE
2 49 Chromatography refrigerator; 49 cu ft, 115V
Call For Price
COMPARE
2 49 Chromatography refrigerator; 49 cu ft, with chart recorder, 115V
Call For Price
COMPARE
3 72 Chromatography refrigerator; 72 cu ft, 115V
Call For Price
COMPARE
3 72 Chromatography refrigerator; 72 cu ft, with chart recorder, 115V
Call For Price
Showing 1- 10 of 10