Spillhyder Universal SM/SMS Sorbent Pads
Coverstock uses spunbond material for excellent durability and virtually no linting
Universal sorbents are ideal for environments where there are many different liquids including cutting oils, lubricants, and coolants. Works with both water-based liquids, oil-based liquids, and more. Spunbond coverstock is abrasion resistant - withstands rigorous abuse from heavy foot traffic. High strength will resist tearing when placed under equipment.
14,100.00INR / Case of 100
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Absorption Color
Availability Pricing
NPS Spilfyter Spilhyder Universal Gray Spill Pads, Medium-Weight, 16" x 18", 100/Case WW-86423-36 Mfr # DS-72
COMPARE
24 gal. (91 L) Gray
₹14,100.00INR / Case of 100