Spilfyter Standard Oil-Only Sorbent Rolls
Standard rolls feature above average absorption, strength, and low-linting
Oil-Only sorbents absorb all oil-based liquids and repel water. Ideal for use with lubricants, fuels, mineral oils, and vegetable oils. Designed for standard use applications.
35,600.00 - ₹39,700.00INR / Bag of 1
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Absorption Description
Availability Pricing
NPS Spilfyter Standard Oil-Only White Spill Pad Roll, Heavy-Weight, 32" x 150', 1/Bag WW-86424-00 Mfr # Z-90
COMPARE
64 gal. (242 L) Standard Oil-Only White Spill Pad Roll, Heavy-Weight, 32" x 150', 1/Bag
₹38,400.00INR / Bag of 1
NPS Spilfyter Standard Oil-Only White Spill Pad Roll, Heavy-Weight, 16" x 150', 2/Bag WW-86424-01 Mfr # Z-91
COMPARE
64 gal. (242 L) Standard Oil-Only White Spill Pad Roll, Heavy-Weight, 16" x 150', 2/Bag
₹39,700.00INR / Bag of 2
NPS Spilfyter Standard Oil-Only White Spill Pad Roll, Medium-Weight, 32" x 150', 1/Bag WW-86424-02 Mfr # Z-97
COMPARE
58 gal. (220 L) Standard Oil-Only White Spill Pad Roll, Medium-Weight, 32" x 150', 1/Bag
₹35,600.00INR / Bag of 1