Spilfyter Premium Universal Sorbent Rolls
Premium rolls feature excellent absorption, strength, and low-linting
Universal sorbents are ideal for environments where there are many different liquids including cutting oils, lubricants, and coolants. Works with both water-based liquids, oil-based liquids, and more. Due to low-linting and high strength, premium sorbents are designed for extended-use applications.
33,200.00 - ₹58,900.00INR / Bag of 2
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Absorption Description
Availability Pricing
NPS Spilfyter Premium Universal Gray Spill Roll, Heavy-Weight, 32" x 150', 1/Bag WW-86423-37 Mfr # GFF-90
COMPARE
64 gal. (242 L) Premium Universal Gray Spill Roll, Heavy-Weight, 32" x 150', 1/Bag
₹47,400.00INR / Bag of 1
NPS Spilfyter Premium Universal Green Spill Roll, Heavy-Weight, 32" x 150', 1/Bag WW-86423-38 Mfr # G-90
COMPARE
64 gal. (242 L) Premium Universal Green Spill Roll, Heavy-Weight, 32" x 150', 1/Bag
₹57,800.00INR / Bag of 1
NPS Spilfyter Premium Universal Gray Spill Roll, Heavy-Weight, 16" x 150', 2/Bag WW-86423-39 Mfr # GFF-91
COMPARE
64 gal. (242 L) Premium Universal Gray Spill Roll, Heavy-Weight, 16" x 150', 2/Bag
₹44,800.00INR / Bag of 2
NPS Spilfyter Premium Universal Green Spill Roll, Heavy-Weight, 16" x 150', 2/Bag WW-86423-40 Mfr # G-91
COMPARE
64 gal. (242 L) Premium Universal Green Spill Roll, Heavy-Weight, 16" x 150', 2/Bag
₹57,800.00INR / Bag of 2
NPS Spilfyter Premium Universal Gray Spill Roll, Medium-Weight, 32" x 150', 1/Bag WW-86423-42 Mfr # GFF-97
COMPARE
50 gal. (189 L) Premium Universal Gray Spill Roll, Medium-Weight, 32" x 150', 1/Bag
₹43,700.00INR / Bag of 1
NPS Spilfyter Premium Universal Gray Spill Roll, Medium-Weight, 16" x 150', 2/Bag WW-86423-43 Mfr # GFF-98
COMPARE
50 gal. (189 L) Premium Universal Gray Spill Roll, Medium-Weight, 16" x 150', 2/Bag
₹33,200.00INR / Bag of 2
NPS Spilfyter Premium Universal Green Spill Roll, Medium-Weight, 16" x 150', 2/Bag WW-86423-44 Mfr # G-98
COMPARE
46 gal. (174 L) Premium Universal Green Spill Roll, Medium-Weight, 16" x 150', 2/Bag
₹52,700.00INR / Bag of 2
NPS Spilfyter Premium Universal Green Spill Roll, Light-Weight, 32" x 300', 1/Bag WW-86423-45 Mfr # G-95
COMPARE
64 gal. (242 L) Premium Universal Green Spill Roll, Light-Weight, 32" x 300', 1/Bag
₹58,900.00INR / Bag of 1
NPS Spilfyter Premium Universal Green Spill Roll, Light-Weight, 16" x 300', 2/Bag WW-86423-46 Mfr # G-96
COMPARE
64 gal. (242 L) Premium Universal Green Spill Roll, Light-Weight, 16" x 300', 2/Bag
₹58,900.00INR / Bag of 2
Showing 1- 9 of 9