This product is now discontinued, View Related Products
Spilfyter Mercury Spill Amalgamation Powder
Convert mercury spill to an amalgam, preventing mercury vapors
Amalgamation powder converts mercury into an amalgam, or paste-like consistency. This prevents mercury vapors from being emitted. Absorbs up to 15 g of mercury/100 g of powder.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
NPS Spilfyter Mercury Amalgamation Powder, 2.5 kg Shaker Bottle WW-86424-57 Mfr # 522500
COMPARE
2500 g shaker bottle
Call For Price
NPS Spilfyter Mercury Amalgamation Powder, 500 g Shaker Bottle WW-86424-58 Mfr # 520500
COMPARE
500 g shaker bottle