Spilfyter Chemical/Wastewater Classifiers
Classify unknown spills in 30 seconds
Convenient strips change color to identify risk type and classify spill for proper treatment. Compare color indicator to laminated, reusable reference chart to classify unknown liquid.
15,300.00 - ₹60,600.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
NPS Spilfyter Chemical Spill Classifier Kit; 6 Tests/Kit WW-86423-00 Mfr # 570001
COMPARE
Chemical Spill Classifier Kit; 6 Tests/Kit
₹15,300.00INR / Each
NPS Spilfyter Wastewater Spill Classifier Kit; 6 Tests/Kit WW-86423-02 Mfr # 580001
COMPARE
Wastewater Spill Classifier Kit; 6 Tests/Kit
₹16,300.00INR / Each
NPS Spilfyter Chemical Classifier Kit, 20 Strips, Gloves, Thread, Chart, Plastic Case WW-86424-16 Mfr # 571020
COMPARE
Chemical Classifier Kit, 20 Strips, Gloves, Thread, Chart, Plastic Case
₹60,600.00INR / Each
NPS Spilfyter Wastewater Classifier Kit, 20 Strips, Gloves, Thread, Chart, Plastic Case WW-86424-18 Mfr # 581020
COMPARE
Wastewater Classifier Kit, 20 Strips, Gloves, Thread, Chart, Plastic Case
₹52,600.00INR / Each