Spex® VapLock™ Waste Funnels
Safely and securely capture waste and hazardous solvents
  • Reduces fumes and harmful vapor exhaust
  • Easily connect to Justrite® Centura safety containers
Use these 4" diameter funnels to safely dispose of liquid laboratory waste. A locking hinged lid minimizes leaks and spills when the funnel is closed. Quick-disconnect fitting on models 12020-64 and -65 allows fast and easy connection to Justrite Centura safety containers. Funnels are constructed of high-density polyethylene (HDPE) and polypropylene (PP) with a Viton® gasket.
30,300.00 - ₹40,700.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Diameter (in) Material Compatible With
Availability Pricing
COMPARE
4 PP / EPDM quick disconnect, HDPE funnel Justrite Centura containers
₹32,700.00INR / Each
COMPARE
4 SS / PTFE quick disconnect, HDPE funnel Justrite Centura containers
₹40,700.00INR / Each
COMPARE
4 HDPE / PP Ports with 1/2" NPT(F)
₹30,300.00INR / Each