Spex® VapLock™ Threaded Plugs, NPT(M)
Designed for secure leak-resistant connections
  • Available in multiple materials and sizes
  • Wrap with PTFE tape for leak-resistant connections
Use plugs to seal unused manifold and bottle cap ports for a secure leak-resistant system. Options for NPT ports are 1/8", 1/4", 3/8", and 1/2" NPT. To ensure a leak-resistant connection, the plugs should be wrapped three to four times with PTFE tape.
60.00 - ₹4,500.00INR / Pkg of 10
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fitting Material Connection Type Thread size NPT(M) (port 1) (in)
Availability Pricing
COMPARE
Polypropylene NPT(M) 1/4
₹2,000.00INR / Pkg of 10
COMPARE
Polypropylene NPT(M) 1/2
₹2,700.00INR / Pkg of 10
COMPARE
PTFE NPT(M) 1/4
₹4,500.00INR / Each
COMPARE
Polypropylene NPT(M) 3/8
₹1,900.00INR / Pkg of 10
COMPARE
Polypropylene NPT(M) 1/8
₹2,100.00INR / Pkg of 10
COMPARE
Polypropylene NPT 1/8
₹60.00INR / Each
COMPARE
Polypropylene NPT(M) 1/4
₹190.00INR / Each
Showing 1- 7 of 7