Spex® VapLock™ Solvent Waste Kits for Justrite® Containers
Collect effluent from multiple sources while minimizing harmful solvent exhaust
  • Easily connect to Justrite safety containers using quick disconnects
Use the waste kits to minimize leaks, spills, and vapor escape from solvent waste containers. Waste kits include a manifold, exhaust filter, adapters, plugs, and tubing—everything you need to install on a typical HPLC instrument. Manifold allows solvent waste to be collected from multiple effluent streams. Exhaust filter minimize exposure to harmful chemical vapors. Fittings and tubing provide leak-tight connections minimizing spills and leaks.
58,000.00 - ₹75,400.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cap / Stopper Size Material Description
Availability Pricing
COMPARE
Quick disconnect Polypropylene Solvent Waste Kit, PP, Justrite PP, 1x1/2"NPTF, 3x1/4"NPTF, 6x1/4"-28 UNFF; 1/ea
₹58,000.00INR / Each
COMPARE
Quick disconnect Polypropylene Solvent Waste Kit, PP, Justrite SS, 1x1/2"NPTF, 3x1/4"NPTF, 6x1/4"-28 UNFF; 1/ea
₹68,900.00INR / Each
COMPARE
Quick disconnect PTFE Solvent Waste Kit, PTFE, Justrite SS, 1x1/2"NPTF, 3x1/4"NPTF, 6x1/4-28UNFF; 1/ea
₹75,400.00INR / Each