Spex® VapLock™ Solvent Waste Kits for Drums and Panel Mounting
Collect effluent from multiple sources while minimizing harmful solvent exhaust
  • Various cap sizes available to fit your solvent container
Use the waste kits to minimize leaks, spills, and vapor escape from solvent waste containers. Waste kits include a manifold, exhaust filter, adapters, plugs, and tubing—everything you need to install on a typical HPLC instrument. Manifold allows solvent waste to be collected from multiple effluent streams. Exhaust filter minimize exposure to harmful chemical vapors. Fittings and tubing provide leak-tight connections minimizing spills and leaks.
57,600.00 - ₹72,000.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cap / Stopper Size Material
Availability Pricing
COMPARE
2" drum bung with fine threads Polypropylene
₹58,200.00INR / Each
COMPARE
2" drum bung with fine threads PTFE
₹69,600.00INR / Each
COMPARE
2" drum bung with coarse threads Polypropylene
₹58,200.00INR / Each
COMPARE
Panel mount Polypropylene
₹57,600.00INR / Each
COMPARE
Panel mount PTFE
₹72,000.00INR / Each