Cole-Parmer® VapLock™ Solvent Collection Systems with Quick Disconnects and FM-Approved Container
Complete solution to safely collect solvent waste from laboratory instruments
  • FM-approved container offers added safety
  • Collect solvent waste from up to 10 effluent streams
  • Minimize exposure to harmful chemical vapors
Solvent waste systems come complete with everything you need to safely collect solvent waste from any system that generates solvent waste. Closed-system design reduces harmful and flammable solvent vapor emissions into the environment as well as leaks and spills.
Systems come with an FM-approved, OSHA, NFPA, and EPA compliant 20-L polyethylene (PE) safety container with quick-disconnect connections plus multiple safety features. A pressure relief lid automatically vents between 3 and 5 psi (0.2 and 0.35 bar) reducing any risk of fire, and a stainless steel arrester prevents flashback ignition. An integral valve prevents vapor release and leaks. A large, self-closing opening and an easy-carry handle with fixed-position latch keeps lid open for convenient pouring. Select from PP or PTFE manifold with quick disconnect fitting to connect up to 10 waste lines.
163,000.00 - ₹288,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (L) Cap / Stopper Size Material
Availability Pricing
COMPARE
20 Quick disconnect Polypropylene(PP)
₹163,000.00INR / Each
COMPARE
20 Quick disconnect Polytetrafluoroethylene (PTFE)
₹288,000.00INR / Each