Spex Vaplock™ Solenoid-Operated Isolation Valves
Fully isolated solenoid allows use with aggressive and high-purity fluids
  • Designed for over 20 million continuous cycles
  • Compact design—ideal for tight spaces
  • Fast response time
  • Very low dead space
Easily dispense precise, repeatable volume using solenoid operated isolation valves. Valves feature a diaphragm that separates the flow path from the operating mechanism ensuring an inert fluid path. Specifically designed to fit in tight spaces, compact valves are great for placing within instruments. Small overall size helps keep the dead volume to a minimum. Fast response time of less than 20 ms ensures flow can be turned on or off in an instant.
31,300.00 - ₹41,700.00INR / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Flow Pattern Orifice (mm) Power (VDC)
Availability Pricing
Spex VapLock™ 2-Way Isolation Valve, Normally Closed, 0.032" Orifice, PTFE Body, PTFE Diaphragm, 20 psi, 19 µL Internal Volume, with Mounting Holes, 12 VDC WW-12021-45 Mfr # 075T2NC12-32M
COMPARE
2-way, normally closed 0.8128 12
₹32,800.00INR / Each
COMPARE
2-way, normally closed 0.8128 24
₹32,800.00INR / Each
Spex VapLock™ 2-Way Isolation Valve, Normally Closed, 0.062" Orifice, PTFE Body, PTFE Diaphragm, 20 psi, 54 µL Internal Volume, 24 VDC WW-12021-47 Mfr # 075T2NC24-62
COMPARE
2-way, normally closed 1.5748 24
₹31,300.00INR / Each
Spex VapLock™ 3-Way Isolation Valve, 0.046" Orifice, PTFE Body, PTFE Diaphragm, 15 psi, 52 µL Internal Volume, 24 VDC WW-12021-51 Mfr # 075T3MP24-46
COMPARE
3-way 1.1684 24
₹41,700.00INR / Each
Spex VapLock™ 2-Way Isolation Valve, Normally Closed, 0.062" Orifice, PTFE Body, PTFE Diaphragm, 30 psi, 55 µL Internal Volume, 12 VDC WW-12021-86 Mfr # 100T2NC12-62
COMPARE
2-way, normally closed 1.5748 12
₹36,400.00INR / Each
Spex VapLock™ 2-Way Isolation Valve, Normally Closed, 0.125" Orifice, PTFE Body, PTFE Diaphragm, 10 psi, 296 µL Internal Volume, 24 VDC WW-12021-87 Mfr # 100T2NC24-125
COMPARE
2-way, normally closed 3.175 24
₹36,400.00INR / Each
Spex VapLock™ 2-Way Isolation Valve, Normally Closed, 0.062" Orifice, PTFE Body, PTFE Diaphragm, 30 psi, 55 µL Internal Volume, 24 VDC WW-12021-88 Mfr # 100T2NC24-62
COMPARE
2-way, normally closed 1.5748 24
₹36,400.00INR / Each
Spex VapLock™ 2-Way Isolation Valve, Normally Open, 0.062" Orifice, PTFE Body, PTFE Diaphragm, 30 psi, 55 µL Internal Volume, 24 VDC WW-12021-90 Mfr # 100T2NO24-62
COMPARE
2-way, normally open 1.5748 24
₹37,900.00INR / Each
Spex VapLock™ 3-Way Isolation Valve, 0.062" Orifice, PTFE Body, PTFE Diaphragm, 30 psi, 95 µL Internal Volume, 24 VDC WW-12021-92 Mfr # 100T3MP24-62
COMPARE
3-way 1.5748 24
₹40,200.00INR / Each
Spex VapLock™ 3-Way Isolation Valve, 0.062" Orifice, PEEK Body, PTFE Diaphragm, 30 psi, 95 µL Internal Volume, 24 VDC WW-12021-93 Mfr # 100T3MP24-62-5
COMPARE
3-way 1.5748 24
₹34,100.00INR / Each
Showing 1- 10 of 10