Cole-Parmer® VapLock™ Reversible Valves for Solvent Management
Maintain pressure equilibrium within the bottle for safe solvent handling
  • Valves feature 0.1 PSI breaking pressure
Valves minimize evaporation and relieve pressure to ensure safe delivery and containment of solvents. Reversible valves serve either purpose depending on the orientation of the valve.
6,700.00 - ₹7,700.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Breaking Pressure (PSI) Connection Type Wetted Materials
Availability Pricing
COMPARE
0.1 1/4\" NPT(M) x 1/4\" NPT(M) PP/EPDM
₹7,000.00INR / Each
COMPARE
0.1 1/4\" NPT(M) x 1/4\" NPT(M) PP/Viton®
₹6,700.00INR / Each
COMPARE
0.1 1/4\" NPT(M) x 1/4\" NPT(M) PP/FFKM
₹7,700.00INR / Each