Cole-Parmer® VapLock™ Relief Valves for Solvent Management
Maintain pressure equilibrium within the bottle for safe solvent handling
  • Choice of 0.1 to 0.25 psi or 1 to 2.5 psi cracking pressure
Valves minimize evaporation and relieve pressure to ensure safe delivery and containment of solvents. Relief valves let pressurized bottle contents escape above the cracking pressure.
3,800.00 - ₹9,300.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Breaking Pressure (PSI) Connection Type Wetted Materials
Availability Pricing
COMPARE
0.1 1/4\"-28 UNF(M) PTFE/Viton®
₹5,000.00INR / Each
COMPARE
0.1 1/4\"-28 UNF(M) PTFE/EPDM
₹5,000.00INR / Each
COMPARE
1 1/4\"-28 UNF(M) PTFE/Viton®
₹5,200.00INR / Each
COMPARE
1 1/4\"-28 UNF(M) PTFE/EPDM
₹3,800.00INR / Each
COMPARE
0.1 1/4\"-28 UNF(M) PTFE/FFKM
₹9,300.00INR / Each
COMPARE
1 1/4\"-28 UNF(M) PTFE/FFKM
₹7,800.00INR / Each