Cole-Parmer® VapLock™ Luer Adapters
Quick connection and release make these luer fittings excellent for many applications
  • Great for connecting to syringes and small tubing
Choose from fittings constructed from durable polypropylene (PP) or chemically inert PVDF.
100.00 - ₹4,000.00INR / Pkg of 10
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connection Type-Port 1 Connection Type-Port 2 Material
Availability Pricing
Cole-Parmer VapLock™ Luer Fitting, PP, Straight, Male Luer Lock x 1/16" Hose Barb; 1/EA WW-12028-46 Mfr # MTLL210-6005
COMPARE
Straight male luer lock 1/16\" hose barb Polypropylene (PP)
₹130.00INR / Each
COMPARE
Straight male luer lock 1/16\" hose barb Polypropylene (PP)
₹4,000.00INR / Pkg of 10
Cole-Parmer VapLock™ Luer Fitting, PP, Straight, Male Luer Lock x 1/8" Hose Barb; 1/EA WW-12028-50 Mfr # MTLL230-6005
COMPARE
Straight male luer lock 1/8\" hose barb Polypropylene (PP)
₹100.00INR / Each
COMPARE
Straight male luer lock 1/8\" hose barb Polypropylene (PP)
₹910.00INR / Pkg of 10
COMPARE
Straight female luer, bulkhead 1/4-28 UNF(M) bottom sealing thread Polypropylene (PP)
₹3,000.00INR / Pkg of 10