Cole-Parmer® VapLock™ Inlet Check Valves for Solvent Management
Maintain pressure equilibrium within the bottle for safe solvent handling
  • Valves feature 0.1 PSI breaking pressure
Valves minimize evaporation and relieve pressure to ensure safe delivery and containment of solvents. Air inlet check valves allow air to enter a bottle to displace vacuum while solvent is pumped out to an instrument.
5,000.00 - ₹7,800.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Breaking Pressure (PSI) Connection Type Wetted Materials
Availability Pricing
COMPARE
0.1 1/4\"-28 UNF(M) PTFE/Viton®
₹5,000.00INR / Each
COMPARE
0.1 1/4\"-28 UNF(M) PTFE/EPDM
₹5,200.00INR / Each
COMPARE
0.1 1/4\"-28 UNF(M) PTFE/FFKM
₹7,800.00INR / Each