Spex® VapLock™ Adapters for Exhaust Filters, NPT(M)
Various adapter configurations allow quick and easy connection to emission sources
VapLock exhaust filters have a 1/4" NPT(F) port and may require an adapter to work with your container. These adapters are NPT(M) x NPT(M), including options for different sizes and choices between straight or elbow.
200.00 - ₹1,900.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fitting Material Connection Type Thread size NPT(M) (port 1) (in)
Availability Pricing
COMPARE
Polypropylene NPT(M) x NPT(M) 1/4
₹200.00INR / Each
COMPARE
Polypropylene NPT(M) x NPT(M) 1/2
₹320.00INR / Each
COMPARE
Polypropylene NPT(M) x NPT(M), 90° elbow 1/2
₹1,900.00INR / Each
COMPARE
Nylon NPT(M) x NPT(M), 45° elbow 1/4
₹1,800.00INR / Each
COMPARE
Polypropylene NPT(M) x NPT(M), 90° elbow 1/4
₹1,500.00INR / Each
COMPARE
Polypropylene NPT(M) x NPT(M) 1/8
₹260.00INR / Each