Cole-Parmer® VapLock™ Adapter Fittings, Hose Barb to Threaded NPT(M), Straight, for Flexible Tubing
Designed for secure leak-resistant connections
  • Constructed of durable polypropylene
Adapters for flexible tubing let you connect 1/16" to 3/4" ID soft-wall tubing to NPT(F) ports. Adapters are made of polypropylene. Wrap NPT threads 3 to 4 times with PTFE tape and wrench tighten for leak-resistant connections.
140.00 - ₹1,300.00INR / Each
21 variations of this product are available.
See All Product Options Below
21 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Fitting Material Thread size NPT(M) (port 2) (in)
Availability Pricing
COMPARE
1/8 Polypropylene 1/4
₹1,000.00INR / Pkg of 10
COMPARE
1/4 Polypropylene 1/4
₹1,000.00INR / Pkg of 10
COMPARE
5/16 Polypropylene 1/4
₹1,200.00INR / Pkg of 10
COMPARE
3/8 Polypropylene 1/4
₹1,100.00INR / Pkg of 10
COMPARE
1/2 Polypropylene 1/4
₹1,300.00INR / Pkg of 10
COMPARE
5/32 Polypropylene 1/4
₹970.00INR / Pkg of 10
COMPARE
1/8 Polypropylene 1/8
₹1,100.00INR / Pkg of 10
COMPARE
1/4 Polypropylene 1/8
₹1,200.00INR / Pkg of 10
COMPARE
5/16 Polypropylene 1/8
₹1,000.00INR / Pkg of 10
COMPARE
3/8 Polypropylene 1/8
₹960.00INR / Pkg of 10
COMPARE
1/4 Polypropylene 1/2
₹920.00INR / Pkg of 10
COMPARE
3/8 Polypropylene 1/2
₹1,200.00INR / Pkg of 10
COMPARE
5/8 Polypropylene 1/2
₹1,200.00INR / Pkg of 10
COMPARE
3/4 Polypropylene 1/2
₹1,300.00INR / Pkg of 10
COMPARE
5/32 Polypropylene 1/4
₹140.00INR / Each
COMPARE
1/8 Polypropylene 1/8
₹180.00INR / Each
COMPARE
1/4 Polypropylene 1/8
₹200.00INR / Each
COMPARE
5/16 Polypropylene 1/8
₹140.00INR / Each
COMPARE
3/8 Polypropylene 1/8
₹170.00INR / Each
COMPARE
5/8 Polypropylene 1/2
₹170.00INR / Each
Showing 1 - 20 of 21