Cole-Parmer® Spex® XRF X-Cell Sample Cup Accessories
Enhance the performance of your XRF X-Cell sample cup
  • Compatible with 40 mm closed X-Cell
  • Compatible with normal optics XRF spectrometers
  • Forms inverted-cone shelf
Use the bubble-free insert with the 3527 04576-05 40 mm XRF liquid cell to prevent trapped air from forming a bubble under the window film when the cell is upright. The insert is pushed into the cell before the sample is added and forms an inverted-cone shelf under which air bubbles can be trapped once the cell is filled and sealed. To use, simply push the insert into the X-Cell after it is partially filled. It is designed to fit into the major suppliers of normal optics XRF spectrometers made by Diano, Rigaku, and Shimadzu.
14,800.00 - ₹125,000.00INR / Pkg of 100
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
40 mm Bubble Free Insert for 04576-05; 100/PK
₹14,800.00INR / Pkg of 100
COMPARE
40 mm Bubble Free Insert for 04576-05; 1000/PK
₹125,000.00INR / Pkg of 1000