Cole-Parmer® Spex® Preloaded Vial Sets for MiniG® and Geno/Grinder® Homogenizers
Preloaded with steel grinding balls for fast and convenient sample preparation
  • Optimize performance of your HG-400 MiniG® and Geno/Grinder® homogenizers
The 4, 5, and 15 mL vial sets come in a foam vial holder within a sturdy polyethylene carrier that has the same footprint and height as a deep-well titer plate. Carrier are used in pairs and fits adjustable clamp assembly 04605-08. Run up to four vial sets at a time for high-throughput tissue homogenization and cell lysis. Use frosted PE vials 04576-97 with Cryo-Block 04500-25 to preserve temperature sensitive samples during grinding. Polyethylene is compatible with commonly used reagents such as TRIzol® solution.
10,400.00 - ₹89,000.00INR / Case of 10
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Description Tube Material Volume (mL)
Availability Pricing
COMPARE
Precleaned Preloaded Vial Set, Polycarbonate, 15 mL Short, 5 Vials/Set; 10 Sets/CS Polycarbonate 15
₹79,900.00INR / Case of 10
COMPARE
Preloaded Vial Set, Polycarbonate, 75 mL, 2/PK Polycarbonate 75
₹10,400.00INR / Pkg of 2
COMPARE
Precleaned Preloaded Vial Set, Polycarbonate, 4 mL, 24 Vials/Set; 10 Sets/CS Polycarbonate 4
₹89,000.00INR / Case of 10
COMPARE
Preloaded Vial Set, Polyethylene, 5 mL, 24 Vials/Set; 10 Sets/CS Polyethylene 5
₹51,500.00INR / Case of 10