Cole-Parmer® Spex® Graphite Crucibles
Cost-effective alternative to precious metal crucibles
  • Chemically inert and heat resistant
  • Disposable
Disposable crucibles eliminate the need for time-consuming cleaning procedures and the possibility of cross-sample contamination. Regular-purity crucibles are made of graphite with impurities ranging from 10 to 100 ppm each of Mg, Fe, Al, Si, and Cu. They are well suited for most basic borate fusion applications. High-purity crucibles are made of graphite with total metallic impurity limits of 2 ppm—ideal for trace-level analytical work.
12,400.00 - ₹71,700.00INR / Pkg of 10
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Bottom Outside Diameter (mm) Capacity (mL) Description
Availability Pricing
COMPARE
38.1 8.4 Regular-Purity Graphite Crucible with Conical Bottom, 8.4 mL; 100/PK
₹71,700.00INR / Pkg of 100
COMPARE
38.1 20 High-Purity Graphite Crucible with Flat Bottom, 20 mL; 10/PK
₹38,000.00INR / Pkg of 10
COMPARE
38.1 20 Regular-Purity Graphite Crucible with Flat Bottom, 20 mL; 10/PK
₹29,600.00INR / Pkg of 10
COMPARE
44.4 27 High-Purity Graphite Crucible with Flat Bottom, 27 mL; 10/PK
₹38,300.00INR / Pkg of 10
COMPARE
44.4 27 Regular-Purity Graphite Crucible with Flat Bttom, 27 mL; 10/PK
₹22,900.00INR / Pkg of 10
COMPARE
50.8 32 High-Purity Graphite Crucible with Flat Bottom, 32 mL; 10/PK
₹42,700.00INR / Pkg of 10
COMPARE
57.1 44.5 High-Purity Graphite Crucible with Flat Bottom, 44.5 mL; 10/PK
₹57,600.00INR / Pkg of 10
COMPARE
38.1 8.4 High-Purity Graphite Crucible with Conical Bottom, 8.4 mL; 10/PK
₹12,400.00INR / Pkg of 10
Showing 1- 8 of 8