This product is now discontinued, View Related Products
Spex® SamplePrep DNAmax Isolation Kits
The complete kit solution for DNA extraction of animal and plant tissue
  • Reinforced tubes designed for homogenization through rapid beat beating
  • Silica-based columns capture DNA and allow for its purification
  • Protocol and workflow optimized for Genomax, Geno/Grinder, and GenoLyte
The Spex Genomic DNA Isolation Kits are optimized to work with the Genomax, Geno/Grinder, and GenoLyte and allow for fast and seamless isolation of DNA from a wide variety of animal tissues and cells, including mammalian, poultry, fish, and soft and tough plant tissue.
The kit combines the use of chaotropic salts and detergents to disrupt animal tissues or cells permitting the selective binding of DNA to a silica column. Proteinase K and RNase A are then added to implement DNA isolation. An ethanol wash buffer is used to remove any additional elements or contamination from the silica column. The column is dried by centrifugation and DNA is eluted using the elution buffer.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Material Compatible With Description
Availability Pricing
COMPARE
Animal Tissue Genomax, Geno/Grinder, GenoLyte DNA Isolation Kit for Animal Tissue; 75 Reactions
Call For Price
COMPARE
Animal Tissue Genomax, Geno/Grinder, GenoLyte DNA Isolation Kit for Animal Tissue with Stabilization Buffer; 75 Reactions
Call For Price
COMPARE
Soft plant tissue Genomax, Geno/Grinder, GenoLyte DNA Isolation Kit for Soft Plant Tissue; 75 Reactions
Call For Price
COMPARE
Tough plant tissue Genomax, Geno/Grinder, GenoLyte DNA Isolation Kit for Tough Plant Tissue; 75 Reactions