Cole-Parmer® Spex® Acid Washed Grinding Beads
Treated to remove fine particles and contaminants
  • Choose from silica or zirconia grinding beads to help remove fine particles and contaminant
Using untreated beads which contain impurities can affect homogenization and biological processes. Acid washing silica and zirconia grinding beads help remove fine particles and contaminant for cleaner samples. Select silica beads for general grinding applications; zirconia beads have greater density and offer higher impact power. General-purpose grinding beads are suitable for most basic cell lysis applications. Choose RNase/DNase-free treated molecular biology-grade grinding beads to inactivate contaminating enzymes. Low-binding grinding beads are coated to reduce nonspecific binding of nucleic acids and proteins.
25,400.00 - ₹53,400.00INR / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
Compareitem specifications
Material Particle Size (micron) Description
Availability Pricing
COMPARE
Silica 800 to 1000 Acid Washed Silica Grinding Beads, 800 to 1,000 µm; 200 g Bottle
₹25,400.00INR / Each
COMPARE
Silica 800 to 1000 Acid Washed Silica Grind Beads, Molecular Biology Grade, 800 to 1,000 µm; 200 g Bottle
₹37,500.00INR / Each
COMPARE
Silica 800 Acid Washed Silica Grinding Beads, Low-Binding, 800 µm; 200 g Bottle
₹37,500.00INR / Each
COMPARE
Silica 400 to 600 Acid Washed Silica Grinding Beads, 400 to 600 µm; 200 g Bottle
₹25,400.00INR / Each
COMPARE
Silica 400 to 600 Acid Washed Silica Grind Beads, Molecular Biology Grade, 400 to 600 µm; 200 g Bottle
₹37,500.00INR / Each
COMPARE
Silica 400 Acid Washed Silica Grinding Beads, Low-Binding, 400 µm; 200 g Bottle
₹37,500.00INR / Each
COMPARE
Silica 100 Acid Washed Silica Grinding Beads, Low-Binding, 100 µm; 200 g Bottle
₹37,500.00INR / Each
COMPARE
Zirconia 200 to 400 Acid Washed Zirconia Grinding Beads, Molecular Biology Grade, 200 to 400 µm; 250 g Bottle
₹52,300.00INR / Each
COMPARE
Zirconia 100 Acid Washed Zirconia Grinding Beads, Low-Binding, 100 µm; 250 g Bottle
₹53,400.00INR / Each
COMPARE
Zirconia 200 Acid Washed Zirconia Grinding Beads, Low-Binding, 200 µm; 250 g Bottle
₹52,300.00INR / Each
COMPARE
Zirconia 3000 Acid Washed Zirconia Grinding Beads, 3 mm; 300 g Bottle
₹31,500.00INR / Each
Showing 1- 11 of 11