This product is now discontinued, View Related Products
Speirs Robertson AMFH Series Microscope Breadboard with Active Air Isolation
Reduces up to 99% of vibrations
  • Ferromagnetic satin silicone worktop with M6 x 25 mm hole pattern
  • Air damped rolling diaphragm sub 2 Hz isolators remove vibrations
  • Stainless steel worktop meets stringent cleanliness requirements
  • 1 year
The air damped rolling diaphragm isolators of the breadboards feature a low natural frequency of 2Hz which removes up to 99% of vibrations. A pattern of M6 holes on a 25 mm grid allowing components to be securely fixed to the platform. The use of ferromagnetic stainless allows magnetic clamps to also be used. A modular range of options allows you to configure tables (sold separately) to your own specifications.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Capacity (lbs) Platform Length (in) Platform Width (in)
Availability Pricing
Speirs Robertson AMFH Microscope Breadboard, Fixing Hole Grid; 75x75 cm WW-11500-43 Mfr # AMFH-75X75
COMPARE
220 29 1/2 29 1/2
Call For Price
Speirs Robertson AMFH Microscope Breadboard, Fixing Hole Grid; 90x75 cm WW-11500-44 Mfr # AMFH-90X75
COMPARE
220 35 2/5 29 1/2
Call For Price
Speirs Robertson AMFH Microscope Breadboard, Fixing Hole Grid; 120x75 cm WW-11500-45 Mfr # AMFH-120X75
COMPARE
220 47 1/4 29 1/2