This product is now discontinued, View Related Products
Spectrum Chromatography Columns For Organic Solutions
Column design minimizes dead volume
Spectrum chromatography columns are made from premium borosilicate glass. Each column is constructed with non-clogging bed supports made of a thin, 10-ìm membrane and a 50-ìm woven grid. All columns are autoclavable.
Columns for Organic Solutions provide excellent chemical resistance with only PTFE and borosilicate glass wetted parts. Ideal for use with solvents (except hydrofluoric acid-solutions).
Call For Price
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Total Volume Capacity (mL) Column Volume (mL/cm) Description
Availability Pricing
Spectrum Chromatography Column for Organic Solutions, 9.4 mL Total Volume WW-34697-06 Mfr # 125001
COMPARE
9.4 0.6 Column for Organic Solutions, 9.4 mL Total Volume
Call For Price
Spectrum Chromatography Column for Organic Solutions, 19 mL Total Volume WW-34697-08 Mfr # 125003
COMPARE
19 0.6 Column for Organic Solutions, 19 mL Total Volume
Call For Price
Spectrum Chromatography Column for Organic Solutions, 35 mL Total Volume WW-34697-20 Mfr # 125009
COMPARE
35 1.7 Column for Organic Solutions, 35 mL Total Volume
Call For Price
Spectrum Chromatography 125011 Column for Organic Solutions, 53 mL Total Volume WW-34697-22 Mfr # 125011
COMPARE
53 1.7 Column for Organic Solutions, 53 mL Total Volume
Call For Price
Spectrum Chromatography 125022 Column for Organic Solutions, 98 mL Total Volume WW-34697-36 Mfr # 125022
COMPARE
98 4.9 Column for Organic Solutions, 98 mL Total Volume
Call For Price
Spectrum Chromatography 125023 Column for Organic Solutions, 147 mL Total Volume WW-34697-38 Mfr # 125023
COMPARE
147 4.9 Column for Organic Solutions, 147 mL Total Volume
Call For Price
Spectrum Chromatography 125029 Column for Organic Solutions, 294 mL Total Volume WW-34697-40 Mfr # 125029
COMPARE
294 4.9 Column for Organic Solutions, 294 mL Total Volume
Call For Price
Spectrum Chromatography 125037 Column for Organic Solutions, 588 mL Total Volume WW-34697-50 Mfr # 125037
COMPARE
588 19.6 Column for Organic Solutions, 588 mL Total Volume
Call For Price
Spectrum Chromatography 125039 Column for Organic Solutions, 1176 mL Total Volume WW-34697-52 Mfr # 125039
COMPARE
1176 19.6 Column for Organic Solutions, 1176 mL Total Volume
Call For Price
Spectrum Chromatography Column for Organic Solutions, 1960 mL Total Volume WW-34697-54 Mfr # 125043
COMPARE
1960 19.6 Column for Organic Solutions, 1960 mL Total Volume
Call For Price
Showing 1- 10 of 10