This product is now discontinued, View Related Products
Spectrum Chromatography Columns For Aqueous Solutions
Column design minimizes dead volume
Spectrum chromatography columns are made from premium borosilicate glass. Each column is constructed with non-clogging bed supports made of a thin, 10-ìm membrane and a 50-ìm woven grid. All columns are autoclavable.
Columns For Aqueous Solutions include PTFE, polypropylene, borosilicate glass, and nitrile rubber wetted parts. Ideal for most aqueous solutions—including most acids, bases, and salts.
Call For Price
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Total Volume Capacity (mL) Column Volume (mL/cm) SPE Cartridge Length (cm)
Availability Pricing
Spectrum Chromatography 124000 Column for Aqueous Solutions, 9.4 mL Total Volume WW-34697-00 Mfr # 124000
COMPARE
9.4 0.6 15
Call For Price
Spectrum Chromatography Column for Aqueous Solutions, 19 mL Total Volume WW-34697-02 Mfr # 124002
COMPARE
19 0.6 30
Call For Price
Spectrum Chromatography 124008 Column for Aqueous Solutions, 35 mL Total Volume WW-34697-12 Mfr # 124008
COMPARE
35 1.7 20
Call For Price
Spectrum Chromatography Column for Aqueous Solutions, 53 mL Total Volume WW-34697-14 Mfr # 124010
COMPARE
53 1.7 20
Call For Price
Spectrum Chromatography 124021 Column for Aqueous Solutions, 98 mL Total Volume WW-34697-28 Mfr # 124021
COMPARE
98 4.9 20
Call For Price
Spectrum Chromatography 124022 Column for Aqueous Solutions, 147 mL Total Volume WW-34697-30 Mfr # 124022
COMPARE
147 4.9 30
Call For Price
Spectrum Chromatography 124028 Column for Aqueous Solutions, 294 mL Total Volume WW-34697-32 Mfr # 124028
COMPARE
294 4.9 60
Call For Price
Spectrum Chromatography Column for Aqueous Solutions, 588 mL Total Volume WW-34697-44 Mfr # 124036
COMPARE
588 19.6 30
Call For Price
Spectrum Chromatography 124038 Column for Aqueous Solutions, 1176 mL Total Volume WW-34697-46 Mfr # 124038
COMPARE
1176 19.6 60
Call For Price
Spectrum Chromatography Column for Aqueous Solutions, 1960 mL Total Volume WW-34697-48 Mfr # 124042
COMPARE
1960 19.6 100
Call For Price
Showing 1- 10 of 10