This product is now discontinued, View Related Products
Spectra/Por 5 Regenerated Cellulose (RC) Dialysis Tubing
Available in a variety of diameters and flat widths to meet your application needs
  • Regenerated cellulose (RC) membrane for the gentle separation and purification of solutes
MWCO of 12,000 to 14,000 Dalton
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Flat Width (mm) Max Molecular Weight Cut Off (Mwco) (Dalton) Min Molecular Weight Cut Off (Mwco) (Dalton)
Availability Pricing
Spectra Por S/P 5 5 Standard RC Dialysis Tubing, 12-14 kDalton, 75 mm, 50ft/bx WW-02900-54 Mfr # 132754
COMPARE
75 14000 12000
Call For Price
Spectra Por S/P 5 5 Standard RC Dialysis Tubing, 12-14 kDalton, 140 mm, 50ft/bx WW-02900-56 Mfr # 132757
COMPARE
140 14000 12000