This product is now discontinued, View Related Products

Spectra/Por 4 Regenerated Cellulose (RC) Dialysis Discs and Sheets

Available in a variety of dimensions to meet your application needs
  • Regenerated Cellulose (RC) membrane for the gentle separation and purification of solutes
MWCO of 12,000 to 14,000
Call For Price

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Max Molecular Weight Cut Off (Mwco) (Dalton) Min Molecular Weight Cut Off (Mwco) (Dalton) Description
Availability Pricing
Spectra Por S/P 4 4 RC Dialysis Membrane Disc, 12000-14000 MWCO, 33mm Dia; 50/Pk WW-02890-68 Mfr # 132498
COMPARE
14000 12000 4 RC Dialysis Membrane Disc, 12000-14000 MWCO, 33mm Dia; 50/Pk
Call For Price
Spectra Por S/P 4 4 RC Dialysis Membrane Disc, 12000-14000 MWCO, 47mm Dia; 50/Pk WW-02890-69 Mfr # 132496
COMPARE
14000 12000 4 RC Dialysis Membrane Disc, 12000-14000 MWCO, 47mm Dia; 50/Pk
Call For Price
Spectra Por S/P 4 4 Standard RC Dialysis Flat Disc, 12-14 kDalton, 100 mm dia, 50/pk WW-02890-83 Mfr # 132494
COMPARE
14000 12000 4 Standard RC Dialysis Flat Disc, 12-14 kDalton, 100 mm dia, 50/pk
Call For Price
Spectra Por S/P 4 Spectra/por 4 Membranes Flat sheets; 150 x 150 mm; 25/pack WW-02900-46 Mfr # 132712
COMPARE
14000 12000 Spectra/por 4 Membranes Flat sheets; 150 x 150 mm; 25/pack
Call For Price
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat