Sonicator Replacement Probes with Threaded Tips
Probe selection is determined by sample volume, sample type, and the desired intensity. As tip diameter increases, the intensity/amplitude decreases. Tips erode over time and replacement tips are available separately.
18,300.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Tip Diameter (mm)
Availability Pricing
Qsonica 4406 Ultrasonic Cell Disruptor Replacement Tips, 1/2" dia WW-04711-86 Mfr # 4406
COMPARE
12.7
₹18,300.00INR / Each
Qsonica 407 Ultrasonic Cell Disruptor Replacement Tips, 3/4" dia WW-04711-87 Mfr # 4407
COMPARE
19
₹18,300.00INR / Each
Qsonica 408 Ultrasonic Cell Disruptor Replacement Tips, 1" dia WW-04711-88 Mfr # 4408
COMPARE
25
₹18,300.00INR / Each