Silicone bubbles make these stoppers lightweight—they float on water. Withstand temperatures from –76 to 392°>F (–60 to 200°>C). Stoppers are available in 14 color-coded sizes, given in both standard and Japanese size numbers. Softness and flexibility allow stoppers to fit a range of holes. Easily make openings for glassware fit your exact specifications with the optional cork borer set. 2 bg/cs.*