This product is now discontinued, View Related Products
Snaptwist Scintillation vials
Acheive a leak proof seal without any added pressure during closing
These general-purpose, shoulderless vials can be used for liquid scintillation counting, gamma counting, chromatography, sample storage, and culturing. Available in either opaque high density polyethylene (HDPE) or translucent polypropylene (PP); both feature exclusive Snaptwist closures made of high-flow
polypropylene.
Vials can be securely sealed by simply snapping the caps on; removal of caps requires an easy quarter-turn. A built-in positive lock prevents the cap from popping off, eliminating the danger of spillage associated with other push-on/pull-off caps.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Height (mm) Volume (mL)
Availability Pricing
Simport Snaptwist Scintillation Vials, PE, 1000/CS WW-08918-10 Mfr # S207
COMPARE
57 6.5
Call For Price
Simport Snaptwist Scintillation Vials, PP, 1000/CS WW-08918-15 Mfr # S207-5
COMPARE
57 6.5