This product is now discontinued, View Related Products
SnakeEye Remote Video Inspection Kits
Easily view hard-to-reach places—identify specific problems and their location
  • Heavy-duty construction withstands shocks and vibrations—use in harsh industrial environments.
These lightweight, modular SnakeEye handheld remote video inspection kits can be easily configured to inspect behind walls, inside ceilings, and in and around pipes and machinery. Kits resist water, dust, and dirt particles and can be used where grease and nonsolvent based chemicals are present.
A hand-held display unit combines a full color CCD camera and TFT-LCD display technology that delivers clear, full color video at a fraction of the price of other remote video systems. Display unit features a high resolution (960 x 234), 5'' rotatable, antiglare display screen that weighs only two pounds. A video in/out port at the base of the unit allows connection to a standard VCR to record and play back images (NTSC format).
Interchangeable camera head measures 1 1/4'' diameter and attaches to either the rigid wand, ring finger attachment, gooseneck adapter, or extension cables (order separately). Ring adapter is ideal for fingertip control in difficult to see but easy to reach places. Battery provides up to 90 minutes of continuous operation.
Kits include: display unit, camera head, 18'' rigid wand adapter, ring finger attachment, 4-ft cable, NiMH battery (set of two) and charger; AC, DC and 12 V adapters, 4-ft video I/O cable, and carrying case.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
SnakeEye BK01W Basic Remote Video Inspection Kit WW-48610-00 Mfr # BK01W
COMPARE
Basic Remote Video Inspection Kit
Call For Price
SnakeEye BK01CW Deluxe Remote Video Inspection Kit WW-48610-10 Mfr # BK01CW
COMPARE
Deluxe Remote Video Inspection Kit