Smart Gasket® with the Spore Trap
Solution to prevent loss of Spore Test Strips in process system
  • Designed to securely retain and accurately expose spore challenge test strips for use in steam sterilization when validating
  • Safe to use.
  • Ease of installation.
  • User friendly
After each validation cycle, the spore trap, with its securely retained test strip, can easily be removed and rechallenged.
Available without or with 1 port
6,500.00 - ₹15,400.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Spore Trap Gasket-EPDM, 1 port, 1-1/2"
₹6,500.00INR / Each
COMPARE
Spore Trap Gasket-EPDM, 1 port, 2"
₹6,900.00INR / Each
COMPARE
Spore Trap Gasket-EPDM, 1 port, 3"
₹9,000.00INR / Each
COMPARE
Spore Trap Gasket-EPDM, 1 port, 4"
₹15,400.00INR / Each