Single-Grip Handheld Pail and Drum Mixers
  • Air control valve allows easy speed adjustment
  • Low speed mixing and stalling will not overheat as with an electric motor
  • Stainless steel shaft and impeller
Popular for mixing open and closed containers. Suitable for mixing slurries, mortar, petroleum products, and similar viscous materials.
186,000.00 - ₹189,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Impeller Size (in) Shaft Diameter (in) Shaft Length (in)
Availability Pricing
Neptune Hand-held single-grip mixer; 1/4" dia shaft x 15"L, 2-1/2" dia caged impeller WW-50500-45 Mfr # HH12
COMPARE
2.5 0.25 15
₹186,000.00INR / Each
Neptune Hand-held single-grip mixer; 1/2" dia shaft x 21"L, 5" dia caged impeller WW-50500-47 Mfr # HH10
COMPARE
5 0.5 21
₹189,000.00INR / Each
Neptune Hand-held single-grip mixer; 1/2" dia shaft x 40"L, 5" dia caged impeller WW-50500-49 Mfr # HH55
COMPARE
5 0.5 40
₹189,000.00INR / Each