This product is now discontinued, View Related Products
Single- and Multi-Element Standards
Ensure consistent results-standards are quality checked at four different steps
  • Quality ingredients produce quality standards, eliminating discrepancies in your lab
  • Our single element standards are certified by ICP-MS or ICP-OES to levels below parts-per-billion
  • Certificate of analysis shows trace levels of 68 metals down to picogram levels
  • Manufactured to exact specifications in an environment that is ISO 9001:2008 certified and 17025:2005 accredited as well as ISO Guide 34 certified
  • Special formulations-we can create custom blends in any volume you need, from 50 mL to 100 L
7,100.00 - ₹88,700.00INR / Each
50 variations of this product are available.
See All Product Options Below
50 PRODUCT OPTIONS
Compareitem specifications
Element/Ion Description
Availability Pricing
COMPARE
₹11,100.00INR / Each
COMPARE
₹7,100.00INR / Each
COMPARE
₹11,100.00INR / Each
COMPARE
₹7,100.00INR / Each
COMPARE
₹11,100.00INR / Each
COMPARE
₹7,100.00INR / Each
COMPARE
₹11,100.00INR / Each
COMPARE
₹7,100.00INR / Each
COMPARE
₹11,100.00INR / Each
COMPARE
₹7,700.00INR / Each
COMPARE
₹11,100.00INR / Each
COMPARE
₹7,100.00INR / Each
COMPARE
₹11,100.00INR / Each
COMPARE
₹7,100.00INR / Each
COMPARE
₹11,100.00INR / Each
COMPARE
₹7,100.00INR / Each
COMPARE
₹7,100.00INR / Each
COMPARE
₹7,100.00INR / Each
COMPARE
₹7,100.00INR / Each
COMPARE
₹11,100.00INR / Each
Showing 1 - 20 of 50